Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114116050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972282.1 | XP_027828083.1 | 4 - 5 | 1259 | 106 |
Gene Symbol: CGAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972727.1 | XP_027828528.1 | 4 - 5 | 1199 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |