Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013753.3 | XP_004013802.1 | 2 - 3 | 331 | 63 | |
XM_012187380.3 | XP_012042770.2 | 2 - 3 | 320 | 63 |