Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181884.3 | XP_012037274.1 | 3 - 4 | 910 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |