Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009981.4 | XP_004010030.2 | 6 - 7 | 989 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |