Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017798.4 | XP_004017847.1 | 5 - 6 | 586 | 83 | |
XM_012098586.3 | XP_011953976.1 | 5 - 6 | 670 | 83 |