Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098977.3 | XP_011954367.1 | 5 - 6 | 577 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |