Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101115225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977946.1 | XP_027833747.1 | 1 - 2 | 153 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |