Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010262.4 | XP_004010311.1 | 3 - 4 | 376 | 97 | |
XM_012180830.3 | XP_012036220.1 | 2 - 3 | 232 | 97 | ||
XM_012180831.3 | XP_012036221.1 | 2 - 3 | 1206 | 97 | ||
GenBank mRNA | DQ239677.1 | - | 1 - 2 | 142 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |