Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011142.4 | XP_004011191.2 | 4 - 5 | 355 | 96 | |
GenBank mRNA | FJ712191.1 | - | 3 - 4 | 238 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |