Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102307.2 | XP_014957793.1 | 8 - 9 | 1098 | 83 | |
XM_027958096.1 | XP_027813897.1 | 7 - 8 | 1481 | 83 | ||
XM_027958097.1 | XP_027813898.1 | 9 - 10 | 1173 | 83 | ||
XM_027958098.1 | XP_027813899.1 | 9 - 10 | 1170 | 83 | ||
XM_027958099.1 | XP_027813900.1 | 9 - 10 | 1170 | 83 | ||
XM_027958100.1 | XP_027813901.1 | 8 - 9 | 1301 | 83 | ||
XM_027958101.1 | XP_027813902.1 | 8 - 9 | 1095 | 83 | ||
XM_027958102.1 | XP_027813903.1 | 9 - 10 | 1176 | 83 | ||
XM_027958103.1 | XP_027813904.1 | 8 - 9 | 1099 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |