Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971818.1 | XP_027827619.1 | 13 - 14 | 1978 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |