Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185208.2 | XP_012040598.1 | 3 - 4 | 405 | 89 | |
XM_015098465.2 | XP_014953951.1 | 3 - 4 | 527 | 89 | ||
XM_015098466.2 | XP_014953952.1 | 3 - 4 | 527 | 89 | ||
XM_027975342.1 | XP_027831143.1 | 3 - 4 | 387 | 89 | ||
XM_027975343.1 | XP_027831144.1 | 3 - 4 | 599 | 89 |