Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012099161.3 | XP_011954551.3 | 1 | 1361 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |