Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966308.1 | XP_027822109.1 | 1 - 2 | 417 | 197 | |
XM_027966309.1 | XP_027822110.1 | 1 - 2 | 442 | 197 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |