Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003864.4 | XP_004003913.2 | 5 - 6 | 1863 | 65 | |
XM_015092607.2 | XP_014948093.2 | 5 - 6 | 1469 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |