Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973706.1 | XP_027829507.1 | 2 - 3 | 331 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |