Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107034.3 | XP_011962424.2 | 2 - 3 | 305 | 72 |
Gene Symbol: LOC114118711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979967.1 | XP_027835768.1 | 1 | 278 | 72 |