Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190219.3 | XP_012045609.1 | 10 - 11 | 884 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |