Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190219.3 | XP_012045609.1 | 5 - 6 | 491 | 102 | |
GenBank mRNA | DQ449954.1 | - | 2 - 3 | 135 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |