Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970350.1 | XP_027826151.1 | 3 - 4 | 3231 | 80 | |
XM_027970351.1 | XP_027826152.1 | 3 - 4 | 3230 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |