Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100263.2 | XP_014955749.2 | 6 - 7 | 1094 | 90 | |
XM_027978561.1 | XP_027834362.1 | 6 - 7 | 1206 | 90 | ||
XM_027978562.1 | XP_027834363.1 | 6 - 7 | 1228 | 90 | ||
XM_027978563.1 | XP_027834364.1 | 6 - 7 | 1116 | 90 | ||
XM_027978564.1 | XP_027834365.1 | 6 - 7 | 1191 | 90 | ||
XM_027978565.1 | XP_027834366.1 | 6 - 7 | 1169 | 90 | ||
XM_027978566.1 | XP_027834367.1 | 5 - 6 | 1063 | 90 | ||
XM_027978567.1 | XP_027834368.1 | 5 - 6 | 1254 | 90 | ||
XM_027978568.1 | XP_027834369.1 | 4 - 5 | 783 | 90 | ||
XM_027978569.1 | XP_027834370.1 | 5 - 6 | 872 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |