Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978010.1 | XP_027833811.1 | 4 - 5 | 732 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |