Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094261.2 | XP_014949747.1 | 2 - 3 | 323 | 114 | |
XM_027966593.1 | XP_027822394.1 | 2 - 3 | 302 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |