Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TC2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017950.4 | XP_004017999.1 | 7 - 8 | 925 | 68 | |
XM_027957331.1 | XP_027813132.1 | 7 - 8 | 857 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |