Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974777.1 | XP_027830578.1 | 5 - 6 | 5976 | 83 | |
XM_027974778.1 | XP_027830579.1 | 5 - 6 | 5850 | 83 | ||
XM_027974779.1 | XP_027830580.1 | 3 - 4 | 5760 | 83 | ||
XM_027974780.1 | XP_027830581.1 | 5 - 6 | 6793 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |