Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091953.2 | XP_014947439.2 | 3 - 4 | 1297 | 112 | |
XM_027969491.1 | XP_027825292.1 | 3 - 4 | 378 | 112 | ||
XM_027969500.1 | XP_027825301.1 | 3 - 4 | 2777 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |