Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093523.2 | XP_014949009.2 | 4 - 5 | 857 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |