Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103961.3 | XP_011959351.2 | 3 - 4 | 543 | 130 | |
XM_015104018.2 | XP_014959504.2 | 4 - 5 | 755 | 130 | ||
XM_027961790.1 | XP_027817591.1 | 5 - 6 | 478 | 130 | ||
XM_027961792.1 | XP_027817593.1 | 3 - 4 | 501 | 130 | ||
XM_027961793.1 | XP_027817594.1 | 2 - 3 | 798 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |