Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001823.4 | XP_004001872.2 | 3 - 4 | 627 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |