Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011146.4 | XP_004011195.2 | 4 - 5 | 621 | 84 | |
XM_027972334.1 | XP_027828135.1 | 4 - 5 | 621 | 84 |