Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100526781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012898.4 | XP_004012947.3 | 2 - 3 | 504 | 94 | |
GenBank mRNA | HQ283081.1 | - | 2 - 3 | 482 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |