Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964410.1 | XP_027820211.1 | 2 | 3096 | 110 | |
XM_027964411.1 | XP_027820212.1 | 2 | 2927 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |