Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096372.2 | XP_014951858.1 | 2 | 330 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |