Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101102754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978807.1 | XP_027834608.1 | 3 | 2016 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |