Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099126.2 | XP_014954612.2 | 2 | 2004 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |