Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711756.3 | XP_002711802.1 | 6 - 7 | 998 | 117 | |
XM_008257735.2 | XP_008255957.1 | 6 - 7 | 995 | 117 | ||
GenBank mRNA | AB035256.1 | - | 6 - 7 | 630 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |