Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712035.3 | XP_002712081.1 | 5 - 6 | 802 | 94 | |
GenBank mRNA | AF089895.1 | - | 5 - 6 | 367 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |