Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163069.1 | NP_001156541.1 | 2 | 297 | 57 | |
XM_008258163.2 | XP_008256385.1 | 3 | 384 | 57 | ||
GenBank mRNA | AF203903.1 | - | 1 | 153 | 57 | |
DQ983191.1 | - | 2 | 297 | 57 | ||
JX262378.1 | - | 2 | 326 | 57 | ||
KC461185.1 | - | 3 | 354 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |