Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100008830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708253.3 | XP_002708299.1 | 5 - 6 | 759 | 71 | |
XM_017342290.1 | XP_017197779.1 | 5 - 6 | 759 | 71 | ||
XM_017342293.1 | XP_017197782.1 | 2 - 3 | 367 | 71 | ||
GenBank mRNA | AF458078.1 | - | 1 - 2 | 218 | 71 | |
EF472893.1 | - | 3 - 4 | 509 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |