Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266637.2 | XP_008264859.1 | 8 - 9 | 1243 | 119 | |
XM_008266638.2 | XP_008264860.1 | 7 - 8 | 1098 | 119 | ||
GenBank mRNA | AJ457085.1 | - | 4 - 5 | 387 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |