Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266637.2 | XP_008264859.1 | 9 - 10 | 1374 | 65 | |
XM_008266638.2 | XP_008264860.1 | 8 - 9 | 1229 | 65 | ||
GenBank mRNA | AJ457085.1 | - | 5 - 6 | 518 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus Probe spans exons |