Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266637.2 | XP_008264859.1 | 10 - 11 | 1457 | 128 | |
GenBank mRNA | AJ457085.1 | - | 6 - 7 | 601 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons |