Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721804.2 | XP_002721850.1 | 2346 | 88 | ||
XM_008248810.2 | XP_008247032.1 | 2301 | 88 | |||
XM_008248812.2 | XP_008247034.2 | 462 | 88 | |||
XM_008248816.2 | XP_008247038.1 | 417 | 88 | |||
XM_017337815.1 | XP_017193304.1 | 2349 | 88 | |||
XM_017337816.1 | XP_017193305.1 | 2304 | 88 | |||
GenBank mRNA | AY248719.1 | - | 422 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |