Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266589.1 | XP_008264811.1 | 16 - 17 | 2768 | 84 | |
XM_017346951.1 | XP_017202440.1 | 16 - 17 | 2768 | 84 | ||
GenBank mRNA | AY575210.1 | - | 7 - 8 | 877 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |