Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714413.3 | XP_002714459.1 | 2 - 3 | 365 | 104 | |
GenBank mRNA | D13307.1 | - | 2 - 3 | 334 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Oryctolagus cuniculus Probe spans exons |