Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267819.2 | XP_008266041.2 | 2 - 3 | 312 | 50 | |
GenBank mRNA | EF154428.1 | - | 1 - 2 | 33 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |