Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720606.3 | XP_002720652.1 | 325 | 92 | ||
GenBank mRNA | M96556.1 | - | 236 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |