Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195721.1 | NP_001182650.1 | 5 - 6 | 1005 | 100 | |
XM_008263299.2 | XP_008261521.1 | 6 - 7 | 1484 | 100 | ||
XM_008263300.2 | XP_008261522.1 | 6 - 7 | 1412 | 100 | ||
GenBank mRNA | U04301.1 | - | 5 - 6 | 443 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |