Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082009.1 | NP_001075478.1 | 8 - 9 | 1064 | 54 | |
XM_017343628.1 | XP_017199117.1 | 8 - 9 | 1079 | 54 | ||
XM_017343629.1 | XP_017199118.1 | 8 - 9 | 1079 | 54 | ||
XM_017343630.1 | XP_017199119.1 | 8 - 9 | 1079 | 54 | ||
XM_017343631.1 | XP_017199120.1 | 8 - 9 | 1080 | 54 | ||
XM_017343632.1 | XP_017199121.1 | 8 - 9 | 1080 | 54 | ||
GenBank mRNA | AB050594.1 | - | 8 - 9 | 1064 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |