Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082036.1 | NP_001075505.1 | 1 | 1754 | 80 | |
GenBank mRNA | AF056943.1 | - | 1 | 1754 | 80 | |
AF149787.1 | - | 1 | 1746 | 80 | ||
D45025.1 | - | 1 | 1749 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Inventoried Both primers and probe map within a single exon |